Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AP1867-2-carboxymeth 250mg | 2230613-03-1 | 250 MG
SDP

Catalog No. 5000230039
Encompass_Preferred

AP1867-2-(carboxymethoxy) is a synthetic FKBP12(F36V)-directed ligand moiety used as the FKBP12-binding component in dTAG and PROTAC constructs. Supplied as a high-purity solid for research applications, it serves as a building block for targeted protein degradation synthesis and assay development.

  • High purity solid (99.88%).
  • Molecular weight 693.78 g/mol.
  • Molecular formula C38H47NO11.
  • Suitable for chemical synthesis of dTAG or PROTAC molecules.
  • Available in multiple pack sizes, including 250 mg.

Catalog No. 50-002-30039 Supplier Medchemexpress LLC Supplier No. HY114420250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AP1867-2-(carboxymethoxy) is a synthetic FKBP12(F36V)-directed ligand moiety used as the FKBP12-binding component in dTAG and PROTAC constructs. Supplied as a high-purity solid for research applications, it serves as a building block for targeted protein degradation synthesis and assay development.

  • High purity solid (99.88%).
  • Molecular weight 693.78 g/mol.
  • Molecular formula C38H47NO11.
  • Suitable for chemical synthesis of dTAG or PROTAC molecules.
  • Available in multiple pack sizes, including 250 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.