Learn More
Medchemexpress LLC AP1867-2-carboxymeth 50mg | 2230613-03-1 | 50 MG
SDP
Catalog No. 5000229448
Encompass_Preferred
AP1867-2-(carboxymethoxy) is an AP1867-derived FKBP12-binding building block used in targeted protein degradation chemistry. It functions as a linker-compatible moiety for constructing PROTAC and dTAG molecules and is provided as a high-purity research reagent for medicinal chemistry and chemical biology applications.
  • High purity suitable for research applications.
  • Designed for FKBP12-directed binding in degrader constructs.
  • Compatible with common linker chemistries for conjugation.
  • Available in multiple milligram-scale pack sizes for synthesis.
Catalog No. 50-002-29448 Supplier Medchemexpress LLC Supplier No. HY11442050MG
AP1867-2-(carboxymethoxy) is an AP1867-derived FKBP12-binding building block used in targeted protein degradation chemistry. It functions as a linker-compatible moiety for constructing PROTAC and dTAG molecules and is provided as a high-purity research reagent for medicinal chemistry and chemical biology applications.
  • High purity suitable for research applications.
  • Designed for FKBP12-directed binding in degrader constructs.
  • Compatible with common linker chemistries for conjugation.
  • Available in multiple milligram-scale pack sizes for synthesis.