Learn More
Medchemexpress LLC HY-114420 5mg , AP1867-2-(carboxymethoxy) CAS:2230613-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144205MG
Encompass_Preferred
HY-114420 5mg AP1867-2-(carboxymethoxy) CAS: 2230613-03-1 AP1867-2-(carboxymethoxy), the AP1867 (a synthetic FKBP12F36V-directed ligand) based moiety, binds to CRBN ligand via a linker to form dTAG molecules. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1303