Learn More
Medchemexpress LLC HY-114434 5mg Medchemexpress, AP1867 CAS:195514-23-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144345MG
Encompass_Preferred
Medchemexpress, HY-114434 5mg AP1867 CAS:195514-23-9 AP1867 is a synthetic FKBP12 F36V -directed ligand. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2492