Learn More
Aviva Systems Biology Corp AP1M2 Antibody - N-terminal region (ARP52081_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52081_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AP1M2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human AP1M2. Alternate names: HSMU1B, MU-1B, MU1B, mu2, AP1-mu2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-630