Learn More
Apexbio Technology LLC AP20187, 10mg. Cas: 195514-80-8 MFCD: MFCD28167729. Dimerizer,synthetic and cell-permeable
SDP
Supplier:  Apexbio Technology LLC B127410
Encompass_Preferred
AP20187 is a small dimerizer drug [1].To solve the graft-versus-host disease, the in vivo behavior of the transplanted cells should be controlled. AP20187 is used in the conditional system as a chemical inducer of dimerization (CID). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5212