Learn More
Medchemexpress LLC HY-13992 10mg , AP20187 CAS:195514-80-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1399210MG
Encompass_Preferred
HY-13992 10mg AP20187 CAS: 195514-80-8 AP20187 (B/B Homodimerizer) is a cell-permeable ligand used to dimerize FK506-binding protein (FKBP) fusion proteins and initiate biological signaling cascades and gene expression or disrupt protein-protein interactions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0841