Learn More
Selleck Chemical LLC AP20187-5mg
SDP
Catalog No. 502389688
Encompass_Preferred
AP20187 (B/B Homodimerizer) is a chemical inducer of dimerization that activates FKBP-Casp8.
Catalog No. 50-238-9688 Supplier Selleck Chemical LLC Supplier No. S84875MG
AP20187 (B/B Homodimerizer) is a chemical inducer of dimerization that activates FKBP-Casp8.