Learn More
Electron Microscopy Sciences Omniprobe AP250 Tungsten Probe Tip
SDP
Catalog No. 5033754
Encompass
Custom tip designed from tungsten. Tip radius is < 0.5um with 6 degree taper angle for maximum lifetime service. Compatible with Short-Cut system for direct conversion to TEM grid.
Catalog No. 50-337-54 Supplier Electron Microscopy Sciences Supplier No. 7596005
Custom tip designed from tungsten. Tip radius is < 0.5um with 6 degree taper angle for maximum lifetime service. Compatible with Short-Cut system for direct conversion to TEM grid.