Learn More
Apexbio Technology LLC AP26113, 5mg. Cas: 1197958-12-5 MFCD: MFCD23704187. Anaplastic lymphoma kinase (ALK) inhibitor
SDP
Supplier:  Apexbio Technology LLC A13675
Encompass_Preferred
AP26113 is a novel, synthetic, orally available small-molecule inhibitor of anaplastic lymphoma kinase (ALK), which is a receptor tyrosine kinase in the insulin receptor superfamily, with half maximal inhibitory concentration IC50 ranging from 5 nmol/l to 11 nmol/l. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4694