Learn More
Aviva Systems Biology Corp AP2A2 Antibody - C-terminal region (ARP59974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AP2A2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human AP2A2. Alternate names: ADTAB, CLAPA2, HIP9, HYPJ, HIP-9ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-286-902