Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology AP2M1 (E5X6F) Rabbit Monoclonal Antibody, Human, Mouse, Rat, 100 μL
SDP

Catalog No. 502732094
Encompass

This rabbit monoclonal antibody is designed to detect endogenous levels of total AP2M1 protein. The antibody is produced by immunizing animals with a recombinant protein specific to the carboxy terminus of the human AP2M1 protein. AP2M1 is a 50 kDa subunit of the AP-2 coat assembly protein complex, which is essential for clathrin-mediated endocytosis at the plasma membrane. It functions by recognizing tyrosine-based sorting motifs in cargo proteins to facilitate their incorporation into clathrin-coated pits.

  • Validated application: Western blotting.
  • Species reactivity: Human, mouse, and rat.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide.
  • Recommended dilution for western blotting: 1:1,000.
  • Storage conditions: Store at -20°C. Do not aliquot the antibody.

Catalog No. 50-273-2094 Supplier Cell Signaling Technology Supplier No. 68196S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This rabbit monoclonal antibody is designed to detect endogenous levels of total AP2M1 protein. The antibody is produced by immunizing animals with a recombinant protein specific to the carboxy terminus of the human AP2M1 protein. AP2M1 is a 50 kDa subunit of the AP-2 coat assembly protein complex, which is essential for clathrin-mediated endocytosis at the plasma membrane. It functions by recognizing tyrosine-based sorting motifs in cargo proteins to facilitate their incorporation into clathrin-coated pits.

  • Validated application: Western blotting.
  • Species reactivity: Human, mouse, and rat.
  • Formulated in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and < 0.02% sodium azide.
  • Recommended dilution for western blotting: 1:1,000.
  • Storage conditions: Store at -20°C. Do not aliquot the antibody.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.