Learn More
Medchemexpress LLC AP39 | 1429061-80-2 | 96.4% | 721.77 | C37H38BrO2PS3 | 50MG
SDP
Catalog No. 5000238241
Encompass_Preferred
AP39 is a mitochondria-targeted hydrogen sulfide (H2S) donor used in biomedical research to modulate mitochondrial function and protect cells from oxidative damage. It elevates intracellular H2S levels, helps maintain mitochondrial DNA integrity, and has been investigated in models of ischemia-reperfusion injury and neurodegeneration. The compound is provided as a research-grade solid; follow recommended storage and handling conditions.
Catalog No. 50-002-38241 Supplier Medchemexpress LLC Supplier No. HY12612450MG
AP39 is a mitochondria-targeted hydrogen sulfide (H2S) donor used in biomedical research to modulate mitochondrial function and protect cells from oxidative damage. It elevates intracellular H2S levels, helps maintain mitochondrial DNA integrity, and has been investigated in models of ischemia-reperfusion injury and neurodegeneration. The compound is provided as a research-grade solid; follow recommended storage and handling conditions.