Learn More
Aviva Systems Biology Corp AP3S1 Antibody - C-terminal region (ARP51936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AP3S1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human AP3S1. Alternate names: CLAPS3, Sigma3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-610