Learn More
Andwin Scientific AP96 P20 TIPS NONSTERILE 960PK
SDP
Catalog No. NC9451863
Encompass_Preferred
The Biomek AP96 P20 tips are made exclusively to be suitable with the Biomek 3000, NX, NXP, FX and FXP workstation. The tips are free of DNA, DNase, RNase, PCR, pyrogen, and trace metals. These have a volume capacity of 100 L and are sold in a pack of 960.
Catalog No. NC9451863 Supplier Andwin Scientific Supplier No. 717254
The Biomek AP96 P20 tips are made exclusively to be suitable with the Biomek 3000, NX, NXP, FX and FXP workstation. The tips are free of DNA, DNase, RNase, PCR, pyrogen, and trace metals. These have a volume capacity of 100 L and are sold in a pack of 960.