Learn More
Beckman Coulter Biomek Tips P20 Sterile
Catalog No. NC9730145
Encompass
These (AP96) P20 sterile pipette tips are designed for the Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations. The P20s are made of 100% premium polypropylene and manufactured to ensure each tip is straight, leak-proof and guaranteed free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. Case of 10 racks.
Catalog No. NC9730145 Supplier Beckman Coulter Supplier No. 717255
These (AP96) P20 sterile pipette tips are designed for the Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations. The P20s are made of 100% premium polypropylene and manufactured to ensure each tip is straight, leak-proof and guaranteed free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. Case of 10 racks.