Learn More
Beckman Coulter Biomek P250 Sterile Tips with Filter, Case of 10 Racks
Catalog No. NC9757251
Encompass
These (AP96) P250 sterile pipette tips with filters are designed exclusively for Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations. Added filter tips reduce the possibility of sample cross-contamination from aerosols. Like all our pipette tips, the P250s are made of 100% premium polypropylene. All pipetting tips are manufactured to ensure straight, leak-proof products guaranteed to be free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. Case of 10 racks.
Catalog No. NC9757251 Supplier Beckman Coulter Supplier No. 717253
These (AP96) P250 sterile pipette tips with filters are designed exclusively for Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations. Added filter tips reduce the possibility of sample cross-contamination from aerosols. Like all our pipette tips, the P250s are made of 100% premium polypropylene. All pipetting tips are manufactured to ensure straight, leak-proof products guaranteed to be free of DNA, DNase/RNase, PCR inhibition, pyrogen/endotoxin and trace metals. Case of 10 racks.