Learn More
Hach Company APA6000 Alkalinity Standard 1
Supplier: Hach Company 2696753
APA6000 Alkalinity Standard 1-Zero Standard-1 LThe APA6000 Alkalinity Process analyzer uses five reagents each of which is supplied in a 1L bottle designed to provide reagents for 30 days of continuous use