Learn More
Hach Company APA6000 Cleaning Solution 1L
Supplier:  Hach Company 2697053
Encompass
APA6000 Alkalinity-Cleaning Solution-1 LThe APA6000 Alkalinity Process analyzer uses five reagents each of which is supplied in a 1L bottle designed to provide reagents for 30 days of continuous use
Catalog No. 50-002-56950