Learn More
Hach Company Hach APA6000 Cleaning Solution, 1 L (for APA6000 Analyzers)
Catalog No. NC3232842 Shop All Hach Company Products
Encompass
APA6000 Cleaning Solution is a 1 L cleaning formulation used to clean the system during the Prime and Instrument Clean functions of APA6000 online analyzers for hardness, copper, and ammonia/monochloramine measurements.
  • Cleans the system during prime and instrument clean functions.
  • Suitable for online hardness, copper, and ammonia/monochloramine analyzers.
  • Packaged volume: 1 L.
Catalog No. NC3232842 Supplier Hach Company Supplier No. 2876453
APA6000 Cleaning Solution is a 1 L cleaning formulation used to clean the system during the Prime and Instrument Clean functions of APA6000 online analyzers for hardness, copper, and ammonia/monochloramine measurements.
  • Cleans the system during prime and instrument clean functions.
  • Suitable for online hardness, copper, and ammonia/monochloramine analyzers.
  • Packaged volume: 1 L.