Learn More
Hach Company APA6000 HR Hardness Reagents
Supplier:  Hach Company 6002100
Encompass
Reagent Set for APA6000 High Range Hardness Analyzer Includes Reagent 1 Bulk solution Reagent 2 Calmagite Threthanol Ammonium in powder 0 5 mg and Bulk solution 1L Reagent 3 Bulk solution APA Acid surfactant wask 1L
Catalog No. 50-002-06972