Learn More
Hach Company APA6000 HR Hardness Standards
Supplier:  Hach Company 6002200
Encompass
Standards kit for APA6000 High Range Hardness Includes Standard 1 (0 mg/L - 1L) Standard 2 (1000 mg/L - 1L)
Catalog No. 50-002-06970