Learn More
Medchemexpress LLC Apabetalone | 1044870-39-4 | 99.34% | 370.40 | 100 MG
SDP
Catalog No. 5000393713
Encompass_Preferred
Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators, specifically targeting the second bromodomain (BD2) with high selectivity. It enhances the production of ApoA-I in hepatocytes, leading to an increase in high-density lipoprotein cholesterol (HDL-C). Its mechanism involves binding to bromodomains of the BET family by competing for acetylated lysine binding sites. This epigenetic modulation of ApoA-I production suggests it as a potential therapeutic agent for atherosclerosis.
  • Selective BET transcriptional regulator inhibitor
  • Targets the second bromodomain (BD2)
  • Increases ApoA-I production in liver cells
  • Elevates high-density lipoprotein cholesterol (HDL-C)
  • Acts via an epigenetic mechanism
  • Investigated for atherosclerosis treatment
Catalog No. 50-003-93713 Supplier Medchemexpress LLC Supplier No. HY16652100MG
Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators, specifically targeting the second bromodomain (BD2) with high selectivity. It enhances the production of ApoA-I in hepatocytes, leading to an increase in high-density lipoprotein cholesterol (HDL-C). Its mechanism involves binding to bromodomains of the BET family by competing for acetylated lysine binding sites. This epigenetic modulation of ApoA-I production suggests it as a potential therapeutic agent for atherosclerosis.
  • Selective BET transcriptional regulator inhibitor
  • Targets the second bromodomain (BD2)
  • Increases ApoA-I production in liver cells
  • Elevates high-density lipoprotein cholesterol (HDL-C)
  • Acts via an epigenetic mechanism
  • Investigated for atherosclerosis treatment