Learn More
Medchemexpress LLC HY-16652 10mg , Apabetalone CAS:1044870-39-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1665210MG
Encompass_Preferred
HY-16652 10mg Apabetalone CAS: 1044870-39-4 Apabetalone (RVX-208) is an inhibitor of BET transcriptional regulators with selectivity for the second bromodomain. The IC50s are 87 μM and 0.51 μM for BD1 and BD2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0978