Learn More
Selleck Chemical LLC Apabetalone
SDP
Catalog No. 5000003438
Encompass_Preferred
Apabetalone (RVX-208 RVX-000222) is a potent BET bromodomain inhibitor with IC50 of 0 510 M for BD2 in a cell-free assay about 170-fold selectivity over BD1 Phase 2
Catalog No. 50-000-03438 Supplier Selleck Chemical LLC Supplier No. S7295100MG
Apabetalone (RVX-208 RVX-000222) is a potent BET bromodomain inhibitor with IC50 of 0 510 M for BD2 in a cell-free assay about 170-fold selectivity over BD1 Phase 2