Learn More
Enzo Life Sciences Apaf-1 monoclonal antibody (13F11) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804349C100
Encompass_Preferred
Clone: 13F11. Isotype: IgG2a. Immunogen: Recombinant mouse Apaf-1 (apoptosis protease-activating factor 1) (aa 1-97) containing the N-terminal CARD domain. Species reactivity: Mouse, Rat. Specificity: Recognizes the CARD domain of Apaf-1. Applications: ICC, IP, WB. Purity: Protein G-affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9875