Learn More
Enzo Life Sciences Apaf-1 monoclonal antibody (18H2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804350C100
Encompass_Preferred
Clone: 18H2. Isotype: IgG2a. Immunogen: Recombinant mouse Apaf-1 (apoptosis protease-activating factor-1) (aa 1-97) containing the N-terminal CARD domain. Species reactivity: Human, Mouse. Specificity: Recognizes the CARD domain of Apaf-1. Applications: ELISA, ICC, IP, WB. Formulation: Liquid. Protein G purified antibody in PBS containing 0.02% sodium azide.
Catalog No. 50-200-9876