Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apaf1 protein (human, recombinant, His-tagged) | 96.0% | 20UG
SDP

Catalog No. 5000175463
Encompass_Preferred

Recombinant human Apaf1 protein (residues S104-L415) is supplied as a lyophilized, N-terminal 6xHis-tagged protein expressed in Pichia pastoris for biochemical and structural studies. The product is provided with high purity and includes buffer and storage guidance to support reliable reconstitution and downstream assays.

  • Recombinant human Apaf1 covering residues S104-L415, His-tagged.
  • Expressed in Pichia pastoris and supplied as lyophilized powder.
  • Molecular weight approximately 37.4 kDa.
  • Purity 96.0% as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Reconstitution recommended at concentrations ≥100 μg/mL for stability.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Catalog No. 50-001-75463 Supplier Medchemexpress LLC Supplier No. HYP70050220UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human Apaf1 protein (residues S104-L415) is supplied as a lyophilized, N-terminal 6xHis-tagged protein expressed in Pichia pastoris for biochemical and structural studies. The product is provided with high purity and includes buffer and storage guidance to support reliable reconstitution and downstream assays.

  • Recombinant human Apaf1 covering residues S104-L415, His-tagged.
  • Expressed in Pichia pastoris and supplied as lyophilized powder.
  • Molecular weight approximately 37.4 kDa.
  • Purity 96.0% as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM Tris-HCl, 0.5 M NaCl, 6% trehalose, pH 8.0.
  • Reconstitution recommended at concentrations ≥100 μg/mL for stability.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.