Learn More
Medchemexpress LLC HY-108634 1mg Medchemexpress, Apafant CAS:105219-56-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1086341MG
Encompass_Preferred
Medchemexpress, HY-108634 1mg Apafant CAS:105219-56-5 Apafant (WEB 2086), a potent platelet-activating factor (PAF) antagonist, inhibits PAF binding to human PAF receptors with a Ki of 9.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0749