Learn More
New England Biolabs, Inc. ApaI – 25000 units
SDP
Catalog No. 50811950
Encompass
A restriction endonuclease that recognizes the sequence G_GGCC^C.
Catalog No. 50-811-950 Supplier New England Biolabs, Inc. Supplier No. R0114L
A restriction endonuclease that recognizes the sequence G_GGCC^C.