Learn More
New England Biolabs, Inc. ApaLI – 12500 units
SDP
Supplier:  New England Biolabs, Inc. R0507L
Encompass
A restriction endonuclease that recognizes the sequence G^TGCA_C.
Catalog No. 50-812-150