Learn More
New England Biolabs, Inc. ApaLI – 12500 units
SDP
Catalog No. 50812151
Encompass
A restriction endonuclease that recognizes the sequence G^TGCA_C.
Catalog No. 50-812-151 Supplier New England Biolabs, Inc. Supplier No. R0507M
A restriction endonuclease that recognizes the sequence G^TGCA_C.