Learn More
Medchemexpress LLC HY-16060 10mg Medchemexpress, Apalutamide CAS:956104-40-8 Purity:>98%
SDP
Catalog No. 501969966
Encompass_Preferred
Medchemexpress, HY-16060 10mg Apalutamide CAS:956104-40-8 Apalutamide (ARN-509) is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9966 Supplier Medchemexpress LLC Supplier No. HY1606010MG
Medchemexpress, HY-16060 10mg Apalutamide CAS:956104-40-8 Apalutamide (ARN-509) is a potent and competitive androgen receptor (AR) antagonist, binding AR with an IC50 of 16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.