Learn More
Medchemexpress LLC Apalutamide-13C,d3 | 956104-40-8 | 99.9% | 481.45 | 500 UG
SDP
Catalog No. 5000394854
Encompass_Preferred
Apalutamide-13C,d3 is a 13C- and deuterium labeled Apalutamide, which is a potent and competitive androgen receptor antagonist. It is intended for research use only and serves as a valuable tool for quantitative analysis in drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporated stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules for quantitation
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-94854 Supplier Medchemexpress LLC Supplier No. HY16060S2500UG
Apalutamide-13C,d3 is a 13C- and deuterium labeled Apalutamide, which is a potent and competitive androgen receptor antagonist. It is intended for research use only and serves as a valuable tool for quantitative analysis in drug development.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporated stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules for quantitation
  • Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs