Learn More
Medchemexpress LLC Apalutamide-cooh | 1332391-04-4 | 99.8% | C20H12F4N4O3S | 10MG
SDP
Catalog No. 5000261213
Encompass_Preferred
Apalutamide-COOH is the carboxylic acid metabolite of apalutamide used as a reference standard in biochemical, pharmacokinetic, and analytical research. The compound is documented with CAS 1332391-04-4, formula C20H12F4N4O3S, and molecular weight 464.39; it is typically supplied as a high-purity powder for laboratory assays.
  • High purity suitable for analytical standards.
  • Useful as a reference metabolite in metabolism and PK studies.
  • Applicable for androgen receptor-related biochemical assays.
  • Stable under recommended storage conditions for long-term use.
  • Supplied as a powder for flexible formulation and preparation.
  • Molecular formula and molecular weight provided for documentation.
Catalog No. 50-002-61213 Supplier Medchemexpress LLC Supplier No. HY13659610MG
Apalutamide-COOH is the carboxylic acid metabolite of apalutamide used as a reference standard in biochemical, pharmacokinetic, and analytical research. The compound is documented with CAS 1332391-04-4, formula C20H12F4N4O3S, and molecular weight 464.39; it is typically supplied as a high-purity powder for laboratory assays.
  • High purity suitable for analytical standards.
  • Useful as a reference metabolite in metabolism and PK studies.
  • Applicable for androgen receptor-related biochemical assays.
  • Stable under recommended storage conditions for long-term use.
  • Supplied as a powder for flexible formulation and preparation.
  • Molecular formula and molecular weight provided for documentation.