Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apalutamide-d4 | 1638885-65-0 | MFCD18384392 | 99.9% | 481.46 g/mol | C21H11D4F4N5O2S | 10 MG
SDP

Catalog No. 5000285563
Encompass_Preferred

Apalutamide-d4 is a deuterium-labeled analogue of apalutamide supplied as a 10 mg analytical standard for research and analytical applications.

  • Deuterium-labeled internal standard for mass spectrometry and analytical assays.
  • CAS number 1638885-65-0 for unambiguous identification.
  • Molecular formula C21H11D4F4N5O2S; molecular weight 481.46 g/mol.
  • High purity: 99.9% suitable for analytical workflows.
  • Packaged as a 10 mg quantity appropriate for reference and method development.

Catalog No. 50-002-85563 Supplier Medchemexpress LLC Supplier No. HY16060S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apalutamide-d4 is a deuterium-labeled analogue of apalutamide supplied as a 10 mg analytical standard for research and analytical applications.

  • Deuterium-labeled internal standard for mass spectrometry and analytical assays.
  • CAS number 1638885-65-0 for unambiguous identification.
  • Molecular formula C21H11D4F4N5O2S; molecular weight 481.46 g/mol.
  • High purity: 99.9% suitable for analytical workflows.
  • Packaged as a 10 mg quantity appropriate for reference and method development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.