Learn More
Enzo Life Sciences Apamin (0.5mg). CAS: 24345-16-2
SDP
Supplier:  Enzo Life Sciences ALX151013MC05
Encompass_Preferred
Originally isolated from Apis mellifera. A small conductance Ca2+-activated K+ channel blocker. Formula: C79H131N31O24S4. MW: 2027.3. Purity: ≥95% (HPLC). Solubility: Soluble in 0.05M acetic acid (5mg/ml); slightly soluble in water (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-7758