Learn More
Cayman Chemical Apararenon 10mg
SDP
Catalog No. 502992153
Encompass_Preferred
A non-steroidal mineralocorticoid receptor antagonist (Ki = 0.104 µM for the human receptor); selective for mineralocorticoid receptors over ERα and ERβ and glucocorticoid, progesterone, and androgen receptors (IC50s = >100 µM for all); increases the urinary ratio of sodium to potassium in adrenalectomized rats; inhibits aldosterone- and nephrectomy-induced increases in systolic blood pressure and urinary albumin levels, as well as myocardial necrosis and fibrosis, cardiac arterial degeneration, and renal tubule damage, in a rat model of renal hypertension at 3 or 10 mg/kg per day
Catalog No. 50-299-2153 Supplier Cayman Chemical Supplier No. 3067110 MG
A non-steroidal mineralocorticoid receptor antagonist (Ki = 0.104 µM for the human receptor); selective for mineralocorticoid receptors over ERα and ERβ and glucocorticoid, progesterone, and androgen receptors (IC50s = >100 µM for all); increases the urinary ratio of sodium to potassium in adrenalectomized rats; inhibits aldosterone- and nephrectomy-induced increases in systolic blood pressure and urinary albumin levels, as well as myocardial necrosis and fibrosis, cardiac arterial degeneration, and renal tubule damage, in a rat model of renal hypertension at 3 or 10 mg/kg per day