Learn More
TARGETMOL CHEMICALS INC APARARENONE 25MG
SDP
Catalog No. 502577388
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apararenone (MT-3995) is a receptor antagonist of mineralocorticoid receptor which can be used to treat non-alcoholic steatohepatitis and diabetic nephropathy. purity: 99%
Catalog No. 50-257-7388 Supplier TARGETMOL CHEMICALS INC Supplier No. T1430125MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apararenone (MT-3995) is a receptor antagonist of mineralocorticoid receptor which can be used to treat non-alcoholic steatohepatitis and diabetic nephropathy. purity: 99%