Learn More
Medchemexpress LLC HY-109002 5mg Medchemexpress, Apararenone CAS:945966-46-1 Purity:>98%
SDP
Catalog No. 501968644
Encompass_Preferred
Medchemexpress, HY-109002 5mg Apararenone CAS:945966-46-1 Apararenone (MT-3995) is a novel non-steroidal mineralocorticoid receptor antagonists under development for the treatment of diabetic nephropathies and non-alcoholic steatohepatitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8644 Supplier Medchemexpress LLC Supplier No. HY1090025MG
Medchemexpress, HY-109002 5mg Apararenone CAS:945966-46-1 Apararenone (MT-3995) is a novel non-steroidal mineralocorticoid receptor antagonists under development for the treatment of diabetic nephropathies and non-alcoholic steatohepatitis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.