Learn More
Abcam Apatinib, 25MG
Catalog No. 502671530
Encompass_Preferred
MW 441.9 Da. Potent and selective inhibitor of VEGFR2, IC₅₀ = 1 nm. Orally bioavailable. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1530 Supplier Abcam Supplier No. AB28244225MG
MW 441.9 Da. Potent and selective inhibitor of VEGFR2, IC₅₀ = 1 nm. Orally bioavailable. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement