Learn More
Cayman Chemical ApatInIb-d8 10mg
SDP
Supplier:  Cayman Chemical 3505510 MG
Encompass_Preferred
An internal standard for the quantification of apatinib by GC- or LC-MS
Catalog No. 50-291-3152