Learn More
Selleck Chemical LLC Apatinib YN968D1mesylate-10mg
SDP
Supplier:  Selleck Chemical LLC S222110MG
Encompass_Preferred
Apatinib mesylate (YN968D1, Rivoceranib) is a potent inhibitor of the VEGF signaling pathway with IC50 values of 1 nM, 13 nM, 429 nM and 530 nM for VEGFR-2, Ret (c-Ret), c-Kit and c-Src, respectively. Apatinib mesylate induces both autophagy and apoptosis.
Catalog No. 50-238-9707