Learn More
Medchemexpress LLC Apatorsen | 915443-09-3 | 98.5% | 10MG
SDP
Catalog No. 5000259600
Encompass_Preferred
Apatorsen sodium is a second-generation antisense oligonucleotide that binds Hsp27 mRNA and inhibits Hsp27 protein production. It is supplied as a sequence-modified oligonucleotide (2'-O-(2-methoxyethyl) and phosphorothioate chemistries) in sodium salt form for research applications.
  • Targets Hsp27 mRNA to reduce Hsp27 protein expression.
  • Sequence-modified chemistry enhances nuclease resistance and binding affinity.
  • Supplied as a sodium salt suitable for in vitro research.
  • High reported purity for experimental consistency (~98.5%).
  • Useful for studying Hsp27 biology and antisense therapeutic mechanisms.
Catalog No. 50-002-59600 Supplier Medchemexpress LLC Supplier No. HY14572210MG
Add to Cart
Add to Cart
Apatorsen sodium is a second-generation antisense oligonucleotide that binds Hsp27 mRNA and inhibits Hsp27 protein production. It is supplied as a sequence-modified oligonucleotide (2'-O-(2-methoxyethyl) and phosphorothioate chemistries) in sodium salt form for research applications.
  • Targets Hsp27 mRNA to reduce Hsp27 protein expression.
  • Sequence-modified chemistry enhances nuclease resistance and binding affinity.
  • Supplied as a sodium salt suitable for in vitro research.
  • High reported purity for experimental consistency (~98.5%).
  • Useful for studying Hsp27 biology and antisense therapeutic mechanisms.