Learn More
Aviva Systems Biology Corp APBA3 Antibody - N-terminal region (AVARP13005_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13005_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APBA3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APBA3. Alternate names: MGC:15815, X11L2, mint3ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-62598