Learn More
Aviva Systems Biology Corp Apbb1 Antibody - N-terminal region (ARP55471_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55471_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Apbb1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Fe65, RirApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-041