Learn More
Aviva Systems Biology Corp APBB1IP Antibody - N-terminal region (ARP57312_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57312_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APBB1IP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APBB1IP. Alternate names: INAG1, PREL1, RARP1, RIAMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54038