Learn More
Aviva Systems Biology Corp APBB3 Antibody - N-terminal region (ARP52554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APBB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APBB3. Alternate names: FE65L2, MGC150555, MGC87674, SRAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-730