Learn More
Business Communication Solutions Micronview Airborne Particle Counter 100LPM Model A110
Catalog No. NC2200698
Encompass_Preferred
The MicronView 100LPM Airborne Particle Counter (APC), Model A110 is a portable ISO 21501-4 compliant particle counter with a minimum particle detection size of 0.5um. Sampling parameters can be established, and particle counts viewed on an easy to read 8” LCD capacitive touchscreen display. Sampling can be manually or automatically started, with cumulative, differential or concentration-based counts reported. Up to 20,000 zones and 200,000 locations can be established on the system to meet your particle monitoring needs.•Size channels – 0.5um, 1.0um, 2.0um, 3.0um, 5.0um, 10.0um•Dimensions (HxWxD) – 254 x 204 x 263 mm•Weight - 13.45lbs/6.1kg (without battery) •Data storage – 3.5 GB, >5 million sample records•Up to 4 hours of battery life•316L Stainless Steel Enclosure•Software features for 21 CFR Part 11 compliance
This item is not returnable. View return policy
Catalog No. NC2200698 Supplier Business Communication Solutions Supplier No. MACHA110
Add to Cart
Add to Cart
Request a Quote
The MicronView 100LPM Airborne Particle Counter (APC), Model A110 is a portable ISO 21501-4 compliant particle counter with a minimum particle detection size of 0.5um. Sampling parameters can be established, and particle counts viewed on an easy to read 8” LCD capacitive touchscreen display. Sampling can be manually or automatically started, with cumulative, differential or concentration-based counts reported. Up to 20,000 zones and 200,000 locations can be established on the system to meet your particle monitoring needs.•Size channels – 0.5um, 1.0um, 2.0um, 3.0um, 5.0um, 10.0um•Dimensions (HxWxD) – 254 x 204 x 263 mm•Weight - 13.45lbs/6.1kg (without battery) •Data storage – 3.5 GB, >5 million sample records•Up to 4 hours of battery life•316L Stainless Steel Enclosure•Software features for 21 CFR Part 11 compliance