Learn More
Enterprise Technology Solutions APC #11 Replacement Battery Cartridge (Black)
SDP
Catalog No. 5000172507
Encompass_Preferred
The black #11 Replacement Battery Cartridge from APC is a maintenance-free sealed lead-acid battery with suspended electrolyte. The cartridge is composed of two separate battery cells with an expected battery life of 3 to 5 years. The replacement battery cartridge supports SU1400XLTNET, SU1400RMXLTNET, SU2200, SU2200NET, SU2200RM, SU2200RMNET, SU2200RMXL, SU2200RMXLT, SU2200XL, SU2200XLNET, SU2200XLTNET, SU3000, SU3000NET, SU3000RM, SU3000RMNET, SU24XLBP, SU48XLBP and SU48BP UPS models.
Catalog No. 50-001-72507 Supplier Enterprise Technology Solutions Supplier No. RBC11
The black #11 Replacement Battery Cartridge from APC is a maintenance-free sealed lead-acid battery with suspended electrolyte. The cartridge is composed of two separate battery cells with an expected battery life of 3 to 5 years. The replacement battery cartridge supports SU1400XLTNET, SU1400RMXLTNET, SU2200, SU2200NET, SU2200RM, SU2200RMNET, SU2200RMXL, SU2200RMXLT, SU2200XL, SU2200XLNET, SU2200XLTNET, SU3000, SU3000NET, SU3000RM, SU3000RMNET, SU24XLBP, SU48XLBP and SU48BP UPS models.